Search
Search

ret-1| Gene Symbol: ret-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129554.5 | NP_001123026.1 | 4 - 5 | 8332 | 85 | |
| NM_001380820.1 | NP_001366847.1 | 2 - 3 | 410 | 85 | ||
| NM_074255.7 | NP_506656.2 | 4 - 5 | 8433 | 85 | ||
Set Membership: |
|