Search
Search

nhr-50| Gene Symbol: nhr-50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_074614.8 | NP_507015.3 | 4 - 5 | 714 | 176 | |
| NR_136352.1 | 2 - 3 | 595 | 176 | |||
| NR_136353.1 | 4 - 5 | 646 | 176 | |||
| NR_136354.1 | 4 - 5 | 670 | 176 | |||
Set Membership: |
|