Search
Search

snx-6| Gene Symbol: snx-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269834.2 | NP_001256763.1 | 9 - 10 | 1391 | 68 | |
| NM_001269835.3 | NP_001256764.1 | 8 - 9 | 1099 | 68 | ||
| NM_001269836.3 | NP_001256765.1 | 7 - 8 | 1000 | 68 | ||
| NM_001269837.3 | NP_001256766.1 | 3 - 4 | 355 | 68 | ||
Set Membership: |
|