Search
Search

fre-1| Gene Symbol: fre-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307796.3 | NP_001294725.1 | 6 - 7 | 1141 | 70 | |
| NM_075474.6 | NP_507875.1 | 6 - 7 | 1130 | 70 | ||
| GenBank mRNA | AY079165.1 | - | 6 - 7 | 1120 | 70 | |
Set Membership: |
|