Search
Search

F26F2.7| Gene Symbol: F26F2.7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001368486.2 | NP_001355520.1 | 10 - 11 | 1444 | 75 | |
| NM_001368488.4 | NP_001355522.1 | 10 - 11 | 1453 | 75 | ||
| NM_001383511.1 | 10 - 11 | 1436 | 75 | |||
| NM_075571.5 | NP_507972.1 | 10 - 11 | 1432 | 75 | ||
Set Membership: |
|