Search
Search

elp-1| Gene Symbol: elp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270021.4 | NP_001256950.1 | 11 - 12 | 1638 | 117 | |
| NM_001270022.3 | NP_001256951.1 | 11 - 12 | 1600 | 117 | ||
Set Membership: |
|