Search
Search

tbc-11| Gene Symbol: tbc-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373217.4 | NP_001360743.1 | 8 - 9 | 850 | 70 | |
| NM_001380899.1 | NP_001367822.1 | 8 - 9 | 850 | 70 | ||
| NM_075777.7 | NP_508178.2 | 8 - 9 | 804 | 70 | ||
| NM_075778.5 | NP_508179.2 | 8 - 9 | 857 | 70 | ||
Set Membership: |
|