Search
Search

| Gene Symbol: dhcr-24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001374978.3 | NP_001362099.1 | 5 - 6 | 1211 | 70 | |
| NM_076062.7 | NP_508463.1 | 5 - 6 | 1238 | 70 | ||
| Gene Symbol: F52H2.5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380922.1 | NP_001367883.1 | 5 - 6 | 1240 | 70 | |
Set Membership: |
|