Search
Search

spc-1| Gene Symbol: spc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270070.3 | NP_001256999.1 | 4 - 5 | 2014 | 88 | |
| NM_001270071.5 | NP_001257000.1 | 4 - 5 | 1996 | 88 | ||
Set Membership: |
|