Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: lim-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001351878.2 | NP_001338857.1 | 3 - 4 | 443 | 86 | |
NM_001351879.2 | NP_001338856.1 | 5 | 593 | 86 | ||
NM_076268.2 | NP_508669.1 | 2 - 3 | 592 | 86 | ||
GenBank mRNA | U72348.1 | - | 2 - 3 | 343 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |