Search
Search

lim-4| Gene Symbol: lim-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001351878.2 | NP_001338857.1 | 6 - 7 | 827 | 74 | |
| NM_001351879.2 | NP_001338856.1 | 7 - 8 | 983 | 74 | ||
| NM_076268.2 | NP_508669.1 | 5 - 6 | 1009 | 74 | ||
| GenBank mRNA | U72348.1 | - | 5 - 6 | 760 | 74 | |
Set Membership: |
|