Search
Search

srv-5| Gene Symbol: srv-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029517.3 | NP_001024688.1 | 4 - 5 | 528 | 109 | |
| NM_001029518.3 | NP_001024689.1 | 4 - 5 | 531 | 109 | ||
Set Membership: |
|