Search
Search

R08E3.1| Gene Symbol: R08E3.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001351880.2 | NP_001338800.1 | 42 - 43 | 5321 | 110 | |
| NM_001380951.1 | NP_001367835.1 | 42 - 43 | 5335 | 110 | ||
Set Membership: |
|