Search
Search

pqn-62| Gene Symbol: pqn-62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313445.3 | NP_001300374.1 | 3 - 4 | 1051 | 69 | |
| NM_001313446.4 | NP_001300375.1 | 5 - 6 | 1348 | 69 | ||
| NM_001380960.1 | NP_001368582.1 | 5 - 6 | 1327 | 69 | ||
Set Membership: |
|