Search
Search

vps-41| Gene Symbol: vps-41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380962.1 | NP_001367568.1 | 12 - 13 | 2633 | 89 | |
| NM_001383575.1 | 12 - 13 | 2638 | 89 | |||
| NM_001383576.1 | 9 - 10 | 2403 | 89 | |||
Set Membership: |
|