Search
Search

sto-2| Gene Symbol: sto-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270091.4 | NP_001257020.1 | 2 - 3 | 192 | 83 | |
| NM_001270092.5 | NP_001257021.1 | 2 - 3 | 1903 | 83 | ||
| NM_001270093.4 | NP_001257022.1 | 2 - 3 | 185 | 83 | ||
| NM_001270094.2 | NP_001257023.1 | 1 - 2 | 76 | 83 | ||
Set Membership: |
|