Search
Search

egl-6| Gene Symbol: egl-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270103.2 | NP_001257032.1 | 5 - 6 | 545 | 130 | |
| NM_001270104.3 | NP_001257033.1 | 4 - 5 | 721 | 130 | ||
| GenBank mRNA | EU884394.1 | - | 4 - 5 | 539 | 130 | |
Set Membership: |
|