Search
Search

igcm-2| Gene Symbol: igcm-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001375051.2 | NP_001362087.1 | 4 - 5 | 489 | 66 | |
| NM_001375052.3 | NP_001362088.1 | 3 - 4 | 436 | 66 | ||
Set Membership: |
|