Search
Search

sek-1| Gene Symbol: sek-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076921.7 | NP_509322.2 | 6 - 7 | 1059 | 89 | |
| GenBank mRNA | AB024087.1 | - | 6 - 7 | 1069 | 89 | |
| AB060731.2 | - | 6 - 7 | 1069 | 89 | ||
Set Membership: |
|