Search
Search

dct-1| Gene Symbol: dct-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076938.6 | NP_509339.4 | 2 - 3 | 468 | 62 | |
| NM_076939.5 | NP_509340.2 | 2 - 3 | 461 | 62 | ||
| GenBank mRNA | AF133832.1 | - | 2 - 3 | 449 | 62 | |
Set Membership: |
|