Search
Search

vav-1| Gene Symbol: vav-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047757.3 | NP_001041222.1 | 11 - 12 | 2151 | 79 | |
| NM_001373412.3 | NP_001359918.1 | 12 - 13 | 2307 | 79 | ||
| NM_001373413.3 | NP_001360680.1 | 12 - 13 | 2294 | 79 | ||
| GenBank mRNA | DQ136172.1 | - | 12 - 13 | 2245 | 79 | |
Set Membership: |
|