Search
Search

rrc-1| Gene Symbol: rrc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029512.5 | NP_001024683.1 | 2 - 3 | 296 | 81 | |
| NM_001029513.6 | NP_001024684.1 | 2 - 3 | 316 | 81 | ||
| NM_001270161.4 | NP_001257090.1 | 2 - 3 | 292 | 81 | ||
Set Membership: |
|