Search
Search

mig-15| Gene Symbol: mig-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029800.3 | NP_001024971.1 | 14 - 15 | 3257 | 115 | |
| NM_001029801.5 | NP_001024972.1 | 14 - 15 | 3242 | 115 | ||
| NM_001029802.5 | NP_001024973.1 | 15 - 16 | 3208 | 115 | ||
| NM_001029803.6 | NP_001024974.1 | 15 - 16 | 3219 | 115 | ||
| GenBank mRNA | AF087131.1 | - | 14 - 15 | 3279 | 115 | |
Set Membership: |
|