Search
Search

glo-3| Gene Symbol: glo-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270179.3 | NP_001257108.1 | 12 - 13 | 1761 | 117 | |
| NR_132549.1 | 11 - 12 | 1883 | 117 | |||
| GenBank mRNA | FJ009048.1 | - | 12 - 13 | 1675 | 117 | |
Set Membership: |
|