Search
Search

pek-1| Gene Symbol: pek-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_077511.8 | NP_509912.1 | 9 - 10 | 2368 | 59 | |
| GenBank mRNA | AF193341.1 | - | 9 - 10 | 2292 | 59 | |
| AF435953.1 | - | 9 - 10 | 2292 | 59 | ||
Set Membership: |
|