Search
Search

pqn-18| Gene Symbol: pqn-18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270274.4 | NP_001257203.1 | 6 - 7 | 919 | 127 | |
| NM_001270275.4 | NP_001257204.1 | 5 - 6 | 832 | 127 | ||
| NM_001270276.4 | NP_001257205.1 | 2 - 3 | 391 | 127 | ||
| NM_001381127.1 | NP_001366740.1 | 8 - 9 | 1539 | 127 | ||
Set Membership: |
|