Search
Search

hlb-1| Gene Symbol: hlb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129713.4 | NP_001123185.1 | 11 - 12 | 2546 | 84 | |
| NM_001322647.4 | NP_001309707.1 | 15 | 2592 | 84 | ||
| NM_001322648.4 | NP_001309708.1 | 14 - 15 | 2672 | 84 | ||
| NM_001322649.4 | NP_001309709.1 | 13 - 14 | 2474 | 84 | ||
| NM_001322650.3 | NP_001309710.1 | 13 | 2594 | 84 | ||
| NM_001322653.5 | NP_001309713.1 | 9 - 10 | 2009 | 84 | ||
| NM_001322654.3 | NP_001309714.1 | 10 - 11 | 2135 | 84 | ||
| NM_001322655.3 | NP_001309703.1 | 10 | 2279 | 84 | ||
| NM_001322656.4 | NP_001309704.1 | 10 | 2405 | 84 | ||
| NM_001322657.3 | NP_001309717.1 | 10 - 11 | 1973 | 84 | ||
| NM_001322658.3 | NP_001309718.1 | 10 - 11 | 2099 | 84 | ||
| NM_001322659.3 | NP_001309705.1 | 6 - 7 | 1583 | 84 | ||
| NM_001322660.3 | NP_001309706.1 | 6 - 7 | 1709 | 84 | ||
| NM_001377622.2 | NP_001364846.1 | 12 - 13 | 2396 | 84 | ||
| NM_001383669.1 | 13 - 14 | 2480 | 84 | |||
| NM_001383670.1 | 12 - 13 | 2282 | 84 | |||
| NM_001383671.1 | 13 - 14 | 2495 | 84 | |||
| NM_077910.5 | NP_510311.2 | 11 - 12 | 2428 | 84 | ||
Set Membership: |
|