Search
Search

del-1| Gene Symbol: del-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001368544.2 | NP_001355359.1 | 14 - 15 | 1812 | 151 | |
| NM_001368545.3 | NP_001355470.1 | 15 | 1956 | 151 | ||
| NM_001368546.2 | NP_001355471.1 | 14 - 15 | 1761 | 151 | ||
Set Membership: |
|