Search
Search

irk-3| Gene Symbol: irk-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001330918.4 | NP_001317893.1 | 4 - 5 | 343 | 102 | |
| NM_001330920.4 | NP_001317903.1 | 3 - 4 | 313 | 102 | ||
| NM_001381146.1 | NP_001368607.1 | 4 - 5 | 358 | 102 | ||
| NM_001381147.2 | NP_001366850.1 | 4 - 5 | 402 | 102 | ||
Set Membership: |
|