Search
Search

nhr-3| Gene Symbol: nhr-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078022.7 | NP_510423.1 | 1 - 2 | 153 | 140 | |
| GenBank mRNA | AF083222.1 | - | 1 - 2 | 164 | 140 | |
| AF273769.1 | - | 1 - 2 | 160 | 140 | ||
Set Membership: |
|