Search
Search
| Gene Symbol: AKAP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620110.3 | XP_005620167.1 | 7 - 8 | 1156 | 95 | |
| XM_022418627.1 | XP_022274335.1 | 7 - 8 | 1157 | 95 | ||
| XM_846274.5 | XP_851367.1 | 7 - 8 | 1347 | 95 | ||
Set Membership: |
|