Search
Search
| Gene Symbol: PARP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005638564.3 | XP_005638621.1 | 6 - 7 | 840 | 87 | |
| XM_005638568.3 | XP_005638625.1 | 6 - 7 | 846 | 87 | ||
| XM_005638569.3 | XP_005638626.1 | 5 - 6 | 1092 | 87 | ||
| XM_005638570.3 | XP_005638627.1 | 5 - 6 | 1092 | 87 | ||
| XM_022412796.1 | XP_022268504.1 | 6 - 7 | 1065 | 87 | ||
| XM_022412797.1 | XP_022268505.1 | 6 - 7 | 976 | 87 | ||
| XM_022412798.1 | XP_022268506.1 | 5 - 6 | 1096 | 87 | ||
| XM_022412799.1 | XP_022268507.1 | 5 - 6 | 1093 | 87 | ||
| XM_848003.5 | XP_853096.2 | 5 - 6 | 1093 | 87 | ||
Set Membership: |
|