Search
Search
| Gene Symbol: EPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005631982.3 | XP_005632039.1 | 10 - 11 | 1583 | 80 | |
| XM_022406170.1 | XP_022261878.1 | 11 - 12 | 1770 | 80 | ||
| XM_533350.6 | XP_533350.4 | 11 - 12 | 1774 | 80 | ||
Set Membership: |
|