Search
Search
| Gene Symbol: MDM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003103.2 | NP_001003103.1 | 1 | 294 | 85 | |
| XM_005625450.3 | XP_005625507.1 | 1 | 437 | 85 | ||
| XM_005625451.3 | XP_005625508.1 | 1 | 320 | 85 | ||
| XM_022423982.1 | XP_022279690.1 | 1 | 294 | 85 | ||
| XM_022423984.1 | XP_022279692.1 | 1 | 376 | 85 | ||
| GenBank mRNA | AB031276.1 | - | 1 | 288 | 85 | |
| AF100705.1 | - | 1 | 294 | 85 | ||
| AF322416.2 | - | 1 | 294 | 85 | ||
| AF322417.2 | - | 1 | 381 | 85 | ||
Set Membership: |
|