Search
Search
| Gene Symbol: IL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003301.1 | NP_001003301.1 | 3 - 4 | 364 | 80 | |
| GenBank mRNA | AF275796.1 | - | 3 - 4 | 307 | 80 | |
| U12234.1 | - | 3 - 4 | 364 | 80 | ||
| Z70045.1 | - | 2 - 3 | 288 | 80 | ||
Set Membership: |
|