Search
Search
| Gene Symbol: DYNLT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003001.1 | NP_001003001.1 | 4 - 5 | 277 | 72 | |
| XM_022415467.1 | XP_022271175.1 | 4 - 5 | 344 | 72 | ||
| GenBank mRNA | AF491301.1 | - | 4 - 5 | 277 | 72 | |
Set Membership: |
|