Search
Search
| Gene Symbol: PTGS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003023.2 | NP_001003023.1 | 8 - 9 | 1429 | 75 | |
| XM_005625008.3 | XP_005625065.1 | 8 - 9 | 1567 | 75 | ||
| XM_005625009.3 | XP_005625066.1 | 8 - 9 | 1546 | 75 | ||
| XM_005625010.3 | XP_005625067.1 | 8 - 9 | 1498 | 75 | ||
| XM_005625011.3 | XP_005625068.1 | 9 - 10 | 1430 | 75 | ||
| XM_005625012.3 | XP_005625069.1 | 7 - 8 | 1424 | 75 | ||
| XM_005625014.3 | XP_005625071.1 | 7 - 8 | 1335 | 75 | ||
| XM_022422518.1 | XP_022278226.1 | 8 - 9 | 1247 | 75 | ||
| GenBank mRNA | AF535138.1 | - | 8 - 9 | 1432 | 75 | |
| AF535139.1 | - | 4 - 5 | 819 | 75 | ||
Set Membership: |
|