Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
我們的服務對象
生物技術
生物製藥
CDMO
實驗室診斷
工業與應用科學
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Cf02625033_m1
See other SFRP2 GE Assays ›
Gene Symbol:
SFRP2
Gene Name:
secreted frizzled related protein 2
Gene Aliases:
-
Chromosome Location:
Chr.15: 51541197 - 51548765 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID
Cf02625033_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: SFRP2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001002987.2
NP_001002987.1
1 - 2
516
110
GenBank mRNA
AJ560716.1
-
1 - 2
516
110
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
G-protein coupled receptor
Canis familiaris
Probe spans exons
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
patterning of blood vessels
chondrocyte development
outflow tract morphogenesis
cardiac left ventricle morphogenesis
apoptotic process
cell-cell signaling
positive regulation of cell proliferation
male gonad development
negative regulation of cardiac muscle cell apoptotic process
negative regulation of epithelial to mesenchymal transition
positive regulation of endopeptidase activity
regulation of neuron projection development
collagen fibril organization
positive regulation of cell growth
negative regulation of cell growth
negative regulation of cell migration
negative regulation of BMP signaling pathway
cellular response to extracellular stimulus
positive regulation of peptidyl-serine phosphorylation
positive regulation of cell adhesion mediated by integrin
post-anal tail morphogenesis
response to drug
negative regulation of mesodermal cell fate specification
embryonic digit morphogenesis
positive regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of JUN kinase activity
positive regulation of fat cell differentiation
positive regulation of osteoblast differentiation
positive regulation of angiogenesis
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
digestive tract morphogenesis
negative regulation of epithelial cell proliferation
negative regulation of peptidyl-tyrosine phosphorylation
convergent extension involved in axis elongation
bone morphogenesis
sclerotome development
negative regulation of dermatome development
hematopoietic stem cell proliferation
cellular response to X-ray
planar cell polarity pathway involved in neural tube closure
Wnt signaling pathway involved in somitogenesis
positive regulation of canonical Wnt signaling pathway
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
regulation of midbrain dopaminergic neuron differentiation
regulation of stem cell division
negative regulation of planar cell polarity pathway involved in axis elongation
fibronectin binding
integrin binding
receptor agonist activity
endopeptidase activator activity
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline