Search
Search
| Gene Symbol: KCNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003329.1 | NP_001003329.1 | 1 | 1634 | 92 | |
| XM_005621307.3 | XP_005621364.1 | 3 | 2325 | 91 | ||
| XM_005621308.3 | XP_005621365.1 | 3 | 2359 | 91 | ||
| GenBank mRNA | L19740.1 | - | 1 | 1634 | 92 | |
Set Membership: |
|