Search Thermo Fisher Scientific
Gene Symbol: CD52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003240.1 | NP_001003240.1 | 1 - 2 | 98 | 87 | |
GenBank mRNA | S77412.1 | - | 1 - 2 | 98 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris Probe spans exons |