Search
Search
| Gene Symbol: KITLG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012735.2 | NP_001012753.1 | 7 - 8 | 912 | 122 | |
| XM_022427652.1 | XP_022283360.1 | 6 - 7 | 839 | 122 | ||
| GenBank mRNA | AY094361.1 | - | 6 - 7 | 671 | 122 | |
| S53329.1 | - | 6 - 7 | 718 | 122 | ||
Set Membership: |
|