Search
Search
| Gene Symbol: HGFAC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031815.2 | NP_001026985.1 | 10 - 11 | 1361 | 99 | |
| XM_022415808.1 | XP_022271516.1 | 10 - 11 | 3152 | 99 | ||
| GenBank mRNA | AY532633.1 | - | 10 - 11 | 1362 | 99 | |
Set Membership: |
|