Search
Search
| Gene Symbol: P2RX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113456.1 | NP_001106927.1 | 436 | 117 | ||
| XM_014107713.2 | XP_013963188.1 | 652 | 117 | |||
| GenBank mRNA | AH014135.2 | - | 436 | 117 | ||
| EU334661.1 | - | 436 | 117 | |||
Set Membership: |
|