Search
Search
| Gene Symbol: BACE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284472.1 | NP_001271401.1 | 690 | 62 | ||
| GenBank mRNA | AB793309.1 | - | 942 | 62 | ||
| AB793310.1 | - | 897 | 62 | |||
| AB793311.1 | - | 867 | 62 | |||
| AB793312.1 | - | 735 | 62 | |||
| AB793313.1 | - | 822 | 62 | |||
| AB793314.1 | - | 690 | 62 | |||
| AY926592.1 | - | 228 | 62 | |||
Set Membership: |
|