Search
Search
| Gene Symbol: BIRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048023.1 | NP_001041488.1 | 5 - 6 | 1308 | 105 | |
| XM_005619507.3 | XP_005619564.1 | 6 - 7 | 2725 | 105 | ||
| GenBank mRNA | DQ223014.1 | - | 5 - 6 | 1308 | 105 | |
Set Membership: |
|