Search
Search
| Gene Symbol: FUCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003250.1 | NP_001003250.1 | 7 - 8 | 1278 | 100 | |
| GenBank mRNA | U29765.1 | - | 7 - 8 | 1278 | 100 | |
| X92448.1 | - | 1278 | 100 | |||
Set Membership: |
|