Search
Search
| Gene Symbol: CAV1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003296.1 | NP_001003296.1 | 2 - 3 | 194 | 67 | |
| XM_005628347.3 | XP_005628404.1 | 2 - 3 | 268 | 67 | ||
| XM_014118875.2 | XP_013974350.1 | 2 - 3 | 314 | 67 | ||
| GenBank mRNA | U47060.1 | - | 2 - 3 | 194 | 67 | |
Set Membership: |
|