Search
Search
| Gene Symbol: PKLR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256018.1 | NP_001242947.1 | 10 - 11 | 1573 | 163 | |
| XM_014114980.2 | XP_013970455.1 | 10 - 11 | 1714 | 163 | ||
| XM_022420763.1 | XP_022276471.1 | 9 - 10 | 1509 | 163 | ||
| XM_022420764.1 | XP_022276472.1 | 10 - 11 | 1591 | 163 | ||
| XM_022420765.1 | XP_022276473.1 | 9 - 10 | 1377 | 163 | ||
| XM_022420766.1 | XP_022276474.1 | 11 - 12 | 1704 | 163 | ||
| GenBank mRNA | AH004600.2 | - | 9 - 10 | 1452 | 163 | |
Set Membership: |
|