Search
Search
| Gene Symbol: MED1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014116603.2 | XP_013972078.1 | 1218 | 73 | ||
| XM_022423023.1 | XP_022278731.1 | 1618 | 73 | |||
| XM_858916.5 | XP_864009.2 | 1723 | 73 | |||
Set Membership: |
|