Search
Search
| Gene Symbol: CCDC92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014107800.2 | XP_013963275.1 | 312 | 57 | ||
| XM_014107801.2 | XP_013963276.1 | 311 | 57 | |||
| XM_014107802.2 | XP_013963277.1 | 352 | 57 | |||
Set Membership: |
|